COMH4211A303 Combi-Blocks, Inc.

Methyl 2,3-dichloro-4-hydroxybenzoate

  • MDL number MFCD27947530
  • Cas Number : 219685-78-6
  • Molecular Weight : 221.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4900

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4900
Availability details for
Quantity
  • Availability
  • Add to Cart