COMH4211A306 Combi-Blocks, Inc.

N-Ethylpyrrolidine-2-carboxamide

  • MDL number MFCD09815725
  • Cas Number : 750514-83-1
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4904

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4904
Availability details for
Quantity
  • Availability
  • Add to Cart