COMH4211A30B Combi-Blocks, Inc.

1-[(Pyrrolidin-2-yl)carbonyl]pyrrolidine

  • MDL number MFCD09808245
  • Cas Number : 202990-49-6
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4909

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4909
Availability details for
Quantity
  • Availability
  • Add to Cart