COMH4211A30D Combi-Blocks, Inc.

4-(4-methoxyphenoxy)benzenesulfonic acid

  • MDL number MFCD32065762
  • Cas Number : 609800-53-5
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-methoxyphenoxy)benzenesulfonic acid
Inchi Code 1S/C13H12O5S/c1-17-10-2-4-11(5-3-10)18-12-6-8-13(9-7-12)19(14,15)16/h2-9H,1H3,(H,14,15,16)
Inchi Key JKKUTAGNBZIAGI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4911

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4911
Availability details for
Quantity
  • Availability
  • Add to Cart