COMH4211A30E Combi-Blocks, Inc.

6-Cyclohexyl-1H-pyridin-2-one

  • MDL number MFCD12032625
  • Cas Number : 1159814-04-6
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4915

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4915
Availability details for
Quantity
  • Availability
  • Add to Cart