COMH4211A312 Combi-Blocks, Inc.

Ethyl (2e)-3-[(2-phenylethyl)amino]but-2-enoate

  • MDL number MFCD31581331
  • Cas Number : 75560-96-2
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9090
Availability details for
Quantity
  • Availability
  • Add to Cart