COMH4211A33C Combi-Blocks, Inc.

N-(1-Benzyl-3,5-dimethyl-1h-pyrazol-4-yl)-2-chloroacetamide

  • MDL number MFCD03724682
  • Cas Number : 515829-75-1
  • Molecular Weight : 277.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(1-benzyl-3,5-dimethyl-1H-pyrazol-4-yl)-2-chloroacetamide
Inchi Code 1S/C14H16ClN3O/c1-10-14(16-13(19)8-15)11(2)18(17-10)9-12-6-4-3-5-7-12/h3-7H,8-9H2,1-2H3,(H,16,19)
Inchi Key JIAMCNNLSCPTDV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9097

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9097
Availability details for
Quantity
  • Availability
  • Add to Cart