COMH4211A33E Combi-Blocks, Inc.

tert-Butyl 3-methyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrolo[2,3-b]pyridine-1-carboxylate

  • MDL number MFCD21607514
  • Cas Number : 2096334-73-3
  • Molecular Weight : 358.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1651

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1651
Availability details for
Quantity
  • Availability
  • Add to Cart