COMH4211A344 Combi-Blocks, Inc.

3-(-4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid benzyl ester

  • MDL number MFCD06795666
  • Cas Number : 880157-10-8
  • Molecular Weight : 338.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2118

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2118
Availability details for
Quantity
  • Availability
  • Add to Cart