COMH4211A34B Combi-Blocks, Inc.

4-(5-Amino-1,3,4-thiadiazol-2-yl)phenylboronic acid pinacol ester

  • MDL number MFCD13182079
  • Cas Number : 2377609-43-1
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3224
Availability details for
Quantity
  • Availability
  • Add to Cart