COMH4211A351 Combi-Blocks, Inc.

1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-3-(2,2,2-trifluoroethyl)urea

  • MDL number MFCD22494642
  • Cas Number : 874299-21-5
  • Molecular Weight : 344.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3724

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3724
Availability details for
Quantity
  • Availability
  • Add to Cart