COMH4211A353 Combi-Blocks, Inc.

2-[(piperidin-1-yl)carbonyl]-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD20486614
  • Cas Number : 1314080-45-9
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5061
Availability details for
Quantity
  • Availability
  • Add to Cart