COMH4211A359 Combi-Blocks, Inc.

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinamide

  • MDL number MFCD11878274
  • Cas Number : 1169402-51-0
  • Molecular Weight : 248.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5575
Availability details for
Quantity
  • Availability
  • Add to Cart