COMH4211A35A Combi-Blocks, Inc.

N-[2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetamide

  • MDL number MFCD22494293
  • Cas Number : 1356943-52-6
  • Molecular Weight : 279.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5605

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5605
Availability details for
Quantity
  • Availability
  • Add to Cart