COMH4211A35C Combi-Blocks, Inc.

5-methyl-3-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,2,4-oxadiazole

  • MDL number MFCD20486748
  • Cas Number : 1056456-23-5
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6059
Availability details for
Quantity
  • Availability
  • Add to Cart