COMH4211A35E Combi-Blocks, Inc.

N-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinamide

  • MDL number MFCD15529716
  • Cas Number : 1201644-50-9
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6120

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6120
Availability details for
Quantity
  • Availability
  • Add to Cart