COMH4211A361 Combi-Blocks, Inc.

1-(morpholin-4-yl)-2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone

  • MDL number MFCD18910277
  • Cas Number : 1092563-23-9
  • Molecular Weight : 331.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6944

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6944
Availability details for
Quantity
  • Availability
  • Add to Cart