COMH4211A363 Combi-Blocks, Inc.

2-(4-Methoxyphenyl)-vinylboronic acid pinacol ester

  • MDL number MFCD04038966
  • Cas Number : 149777-83-3
  • Molecular Weight : 260.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-[(E)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethenyl]phenyl ether
Inchi Code 1S/C15H21BO3/c1-14(2)15(3,4)19-16(18-14)11-10-12-6-8-13(17-5)9-7-12/h6-11H,1-5H3/b11-10+
Inchi Key VXSHTICWQKLRMP-ZHACJKMWSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8136
Availability details for
Quantity
  • Availability
  • Add to Cart