COMH4211A366 Combi-Blocks, Inc.

2-(4-bromobutyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD22419948
  • Cas Number : 124215-50-5
  • Molecular Weight : 263.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8593
Availability details for
Quantity
  • Availability
  • Add to Cart