COMH4211A367 Combi-Blocks, Inc.

2,3-Dihydrothieno[3,4-b][1,4]dioxin-5-ylboronic acid, pinacol ester

  • MDL number MFCD08060541
  • Cas Number : 250726-93-3
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9101

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9101
Availability details for
Quantity
  • Availability
  • Add to Cart