COMH4211A36F Combi-Blocks, Inc.

4,6-dibromo-2-methylpyridin-3-ol

  • MDL number MFCD11044314
  • Cas Number : 188923-75-3
  • Molecular Weight : 266.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1834
Availability details for
Quantity
  • Availability
  • Add to Cart