COMH4211A370 Combi-Blocks, Inc.

4-chloro-N-isopropylpyridine-2-carboxamide

  • MDL number MFCD08705679
  • Cas Number : 604813-08-3
  • Molecular Weight : 198.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7191

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7191
Availability details for
Quantity
  • Availability
  • Add to Cart