COMH4211A374 Combi-Blocks, Inc.

5-bromo-3-chloro-2-ethoxypyridine

  • MDL number MFCD17013174
  • Cas Number : 1245648-68-3
  • Molecular Weight : 236.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7909

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7909
Availability details for
Quantity
  • Availability
  • Add to Cart