COMH4211A3C6 Combi-Blocks, Inc.

1-Methyl-1-(3-nitro-2-pyridyl)hydrazine

  • MDL number MFCD00052235
  • Cas Number : 30963-12-3
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1725

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1725
Availability details for
Quantity
  • Availability
  • Add to Cart