COMH4211A3F2 Combi-Blocks, Inc.

Diethyl 1-butylphosphonate

  • MDL number MFCD08445506
  • Cas Number : 2404-75-3
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5326
Availability details for
Quantity
  • Availability
  • Add to Cart