COMH4211A3FA Combi-Blocks, Inc.

Thiomorpholine-3-carboxylic acid ethyl ester, HCl

  • MDL number MFCD00107968
  • Cas Number : 159381-07-4
  • Molecular Weight : 211.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-thiomorpholinecarboxylate hydrochloride
Inchi Code 1S/C7H13NO2S.ClH/c1-2-10-7(9)6-5-11-4-3-8-6;/h6,8H,2-5H2,1H3;1H
Inchi Key QQRCEZBURAQLBQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5935

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5935
Availability details for
Quantity
  • Availability
  • Add to Cart