COMH4211A43A Combi-Blocks, Inc.

4-Hydrazinocarbonyl-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD06795951
  • Cas Number : 161609-80-9
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8622

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8622
Availability details for
Quantity
  • Availability
  • Add to Cart