COMH4211A454 Combi-Blocks, Inc.

6-Formyl-uracil monohydrate

  • MDL number MFCD03002359
  • Cas Number : 36327-91-0
  • Molecular Weight : 158.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3145
Availability details for
Quantity
  • Availability
  • Add to Cart