COMH4211A46A Combi-Blocks, Inc.

4-[(2-Methylpropoxy)methyl]piperidine

  • MDL number MFCD06247827
  • Cas Number : 883530-03-8
  • Molecular Weight : 171.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0004

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0004
Availability details for
Quantity
  • Availability
  • Add to Cart