COMH4211A46B Combi-Blocks, Inc.

1-(3-Methylbenzoyl)piperidine-3-carboxylic acid

  • MDL number MFCD08445089
  • Cas Number : 923676-22-6
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0005
Availability details for
Quantity
  • Availability
  • Add to Cart