COMH4211A46C Combi-Blocks, Inc.

4-(4-Bromo-2-trifluoromethylbenzenesulfonyl)morpholine

  • MDL number MFCD20485105
  • Cas Number : 877383-38-5
  • Molecular Weight : 374.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-((4-bromo-2-(trifluoromethyl)phenyl)sulfonyl)morpholine
Inchi Code 1S/C11H11BrF3NO3S/c12-8-1-2-10(9(7-8)11(13,14)15)20(17,18)16-3-5-19-6-4-16/h1-2,7H,3-6H2
Inchi Key QDTAAKWVHBMKDE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0008
Availability details for
Quantity
  • Availability
  • Add to Cart