COMH4211A46E Combi-Blocks, Inc.

(4-Bromophenyl)(4-hydroxy-4-methylpiperidin-1-yl)methanone

  • MDL number MFCD22993203
  • Cas Number : 1464158-41-5
  • Molecular Weight : 298.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0014
Availability details for
Quantity
  • Availability
  • Add to Cart