COMH4211A46F Combi-Blocks, Inc.

(2-Bromo-4-methyl-benzyl)-cyclopropyl-carbamic acid tert-butyl ester

  • MDL number MFCD31613863
  • Cas Number : 2270906-20-0
  • Molecular Weight : 340.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0016

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0016
Availability details for
Quantity
  • Availability
  • Add to Cart