COMH4211A470 Combi-Blocks, Inc.

1-(3-Fluorobenzoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD08443435
  • Cas Number : 1104518-12-8
  • Molecular Weight : 237.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0017
Availability details for
Quantity
  • Availability
  • Add to Cart