COMH4211A471 Combi-Blocks, Inc.

2-[1-(Cyclohexylmethyl)piperidin-4-yl]ethan-1-ol

  • MDL number MFCD23708534
  • Cas Number : 141430-54-8
  • Molecular Weight : 225.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0019
Availability details for
Quantity
  • Availability
  • Add to Cart