COMH4211A493 Combi-Blocks, Inc.

3-[1-(2,2,2-Trifluoro-ethyl)-piperidin-4-yl]-propionic acid hydrochloride

  • MDL number MFCD31557823
  • Cas Number : 2206608-91-3
  • Molecular Weight : 275.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0020

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0020
Availability details for
Quantity
  • Availability
  • Add to Cart