COMH4211A494 Combi-Blocks, Inc.

1-(Azetidin-3-yl)-4-methoxy-1h-pyrazole

  • MDL number MFCD31583052
  • Cas Number : 1849281-53-3
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0023
Availability details for
Quantity
  • Availability
  • Add to Cart