COMH4211A495 Combi-Blocks, Inc.

1-(3-Bromo-4-methylphenyl)ethan-1-amine

  • MDL number MFCD18710244
  • Cas Number : 90485-17-9
  • Molecular Weight : 214.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0024

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0024
Availability details for
Quantity
  • Availability
  • Add to Cart