COMH4211A496 Combi-Blocks, Inc.

5-Isobutoxy-2-nitro-benzaldehyde

  • MDL number MFCD30830214
  • Cas Number : 154606-15-2
  • Molecular Weight : 223.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0025
Availability details for
Quantity
  • Availability
  • Add to Cart