COMH4211A497 Combi-Blocks, Inc.

1-[3-Methyl-4-(propan-2-yloxy)phenyl]ethan-1-amine

  • MDL number MFCD11053246
  • Cas Number : 105321-40-2
  • Molecular Weight : 193.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0026

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0026
Availability details for
Quantity
  • Availability
  • Add to Cart