COMH4211A498 Combi-Blocks, Inc.

5-Benzyloxy-2-cyclopropylmethoxy-benzoic acid

  • MDL number MFCD31618914
  • Cas Number : 2270907-98-5
  • Molecular Weight : 298.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0027
Availability details for
Quantity
  • Availability
  • Add to Cart