COMH4211A499 Combi-Blocks, Inc.

(R)-N-(Piperidin-3-yl)thiazole-2-carboxamide

  • MDL number MFCD31557820
  • Cas Number : 1643574-20-2
  • Molecular Weight : 211.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0028
Availability details for
Quantity
  • Availability
  • Add to Cart