COMH4211A49B Combi-Blocks, Inc.

3-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazole-1-carbonyl]-amino-propionic acid methyl ester

  • MDL number MFCD31583142
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0031
Availability details for
Quantity
  • Availability
  • Add to Cart