COMH4211A49C Combi-Blocks, Inc.

4-Fluoro-2,n-dimethyl-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzamide

  • MDL number MFCD31579387
  • Molecular Weight : 293.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0034

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0034
Availability details for
Quantity
  • Availability
  • Add to Cart