COMH4211A49D Combi-Blocks, Inc.

1,3-Difluoro-5-(prop-1-en-2-yl)benzene

  • MDL number MFCD11113479
  • Cas Number : 125661-57-6
  • Molecular Weight : 154.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0035

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0035
Availability details for
Quantity
  • Availability
  • Add to Cart