COMH4211A49E Combi-Blocks, Inc.

Prop-2-en-1-yl n-[2-(2-hydroxyethoxy)ethyl]carbamate

  • MDL number MFCD20287039
  • Cas Number : 918637-04-4
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0036
Availability details for
Quantity
  • Availability
  • Add to Cart