COMH4211A4A1 Combi-Blocks, Inc.

1-Phenyl-1,3,8-triazaspiro[4.5]decane-2,4-dione

  • MDL number MFCD22741704
  • Cas Number : 23364-19-4
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0050

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0050
Availability details for
Quantity
  • Availability
  • Add to Cart