COMH4211A4A3 Combi-Blocks, Inc.

1-[(1E)-3-Bromo-1-propen-1-yl]-3,5-difluoro-benzene

  • MDL number MFCD04116063
  • Cas Number : 210113-74-9
  • Molecular Weight : 233.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0053
Availability details for
Quantity
  • Availability
  • Add to Cart