COMH4211A4A4 Combi-Blocks, Inc.

1-(3-Bromophenyl)-3-(cyclopropylmethyl)urea

  • MDL number MFCD13514847
  • Cas Number : 1209975-39-2
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0054

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0054
Availability details for
Quantity
  • Availability
  • Add to Cart