COMH4211A4A5 Combi-Blocks, Inc.

1-(6-Methylpyridin-2-yl)butane-1,3-dione

  • MDL number MFCD09996567
  • Cas Number : 1020037-93-7
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0057

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0057
Availability details for
Quantity
  • Availability
  • Add to Cart